Specifications
pH10 ± 0.1 at 25°C
Absorbance1.00 max at 210 nm, 0.04 max. at 230 nm, 0.005 max. at 280 nm, 0.005 max. at 350 nm
Color5 max. (APHA)
AppearanceClear, colorless liquid
Quantity4 L
PackagingGlass Bottle
Physical FormLiquid
Purity Grade NotesHPLC and Preparative Chromatography
Unit SizeEach