Chromplete™ Water, HPLC/GC, and Spectrometry, Thermo Scientific™
This product ships via ground from US northeast.
Chromplete™ Water, HPLC/GC, and Spectrometry, Thermo Scientific™
Thermo Scientific™

Chromplete™ Water, HPLC/GC, and Spectrometry, Thermo Scientific™

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Water is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Unit Size:
View all
Each
Case of 6
Catalog number T001031000
Price (USD)/ Each
49.00
-
Add to cart
Quantity:
1 L
Unit Size:
Each
Request bulk or custom format
Price (USD)/ Each
49.00
Add to cart
Chromplete™ Water, HPLC/GC, and Spectrometry, Thermo Scientific™
Catalog numberT001031000
Price (USD)/ Each
49.00
-
Add to cart
Specifications
Absorbance≤ 0.01, ≤ 0.005
Chemical Name or MaterialWater
Shelf Life1825 Days
pH5.5-7.0
CAS7732-18-5
View more

Chromplete Water is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
RUO – Research Use Only