Orion™ Star A Series Portable Meter pH Electrode Holder
Orion™ Star A Series Portable Meter pH Electrode Holder
Thermo Scientific™
Orion™ Star A Series Portable Meter pH Electrode Holder
Use the Thermo Scientific™ Orion™ Star A Series Portable Meter pH Electrode Holder with Star A series portable meter armor and sleeve to protect your pH electrode in the field
Have Questions?
Catalog number STARA-ESPH
Price (USD)/ Each
31.46
Special Offer
Online exclusive
Ends: 31-Dec-2024
36.80 
Save 5.34 (15%)
-
Add to cart
Description:
Orion Star A series pH electrode holder for portable meter protective armor
Price (USD)/ Each
31.46
Special Offer
Online exclusive
Ends: 31-Dec-2024
36.80 
Save 5.34 (15%)
Add to cart
Orion™ Star A Series Portable Meter pH Electrode Holder
Catalog numberSTARA-ESPH
Price (USD)/ Each
31.46
Special Offer
Online exclusive
Ends: 31-Dec-2024
36.80 
Save 5.34 (15%)
-
Add to cart
Use the Thermo Scientific™ Orion™ Star A Series Portable Meter pH Electrode Holder with the Star A series portable meter armor (Cat. No. STARA-AR) and storage sleeve (Cat. No. 810017) to protect, transport and store your sensors in the field. This pH electrode holder can be attached to either side of the portable meter armor and holds one pH electrode when placed in the pH electrode storage sleeve.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionOrion Star A series pH electrode holder for portable meter protective armor
TypeMeter Accessories
Recommended AccessoriespH electrode storage sleeve (Cat. No. 810017)
Unit SizeEach