Orion™ Star A Series Portable Meter Hard-sided Field Case
Orion™ Star A Series Portable Meter Hard-sided Field Case
Thermo Scientific™
Orion™ Star A Series Portable Meter Hard-sided Field Case
Use the Thermo Scientific™ Orion™ Star A Series Portable Meter Hard-sided Field Case with Orion™ Star A portable meters to protect, transport and store equipment in the field
Have Questions?
Catalog number STARA-CS
Price (USD)/ Each
155.61
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 26.39 (15%)
-
Add to cart
Description:
Orion Star A series hard-sided field case
Price (USD)/ Each
155.61
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 26.39 (15%)
Add to cart
Orion™ Star A Series Portable Meter Hard-sided Field Case
Catalog numberSTARA-CS
Price (USD)/ Each
155.61
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 26.39 (15%)
-
Add to cart
Use the Thermo Scientific™ Orion™ Star A Series Portable Meter Hard-sided Field Case with the Thermo Scientific™ Orion™ Star A series portable meters (Star A320, Star A220 and Star A120 series) to protect, transport and store your equipment in the field. Field case holds Star A series portable meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeve. The electrodes and probes can remain connected to meter while in the case plus the internal separating door can be removed for cable management.
Hard-sided field case includes cable management system for electrodes.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionOrion Star A series hard-sided field case
TypeMeter Accessories
Unit SizeEach