Exo-Resistant Random Primer
Exo-Resistant Random Primer
Thermo Scientific™

Exo-Resistant Random Primer

Thermo Scientific Exonuclease-Resistant Random Primer is a mixture of single-stranded random oligonucleotides used for highly efficient random priming of variousRead more
Have Questions?
Catalog number SO181
Price (USD)/ 100 µL
83.75
-
Add to cart
Price (USD)/ 100 µL
83.75
Add to cart
Exo-Resistant Random Primer
Catalog numberSO181
Price (USD)/ 100 µL
83.75
-
Add to cart

Thermo Scientific Exonuclease-Resistant Random Primer is a mixture of single-stranded random oligonucleotides used for highly efficient random priming of various DNA synthesis reactions. This primer has two 3'-terminal phosphorothioate (PTO) modifications that are resistant to the 3'→5' exonuclease activity of proofreading DNA polymerases, such as Klenow Fragment and phi29 DNA Polymerase. It also has 5'- and 3'-hydroxyl ends. The Exo-Resistant Random Primer is supplied as a ready-to-use, 20X concentrated aqueous solution.

Features
• 3'-terminal PTO modifications protect from 3'→5' exonuclease degradation and ensure efficient priming of DNA synthesis
• Product delivered in a ready-to-use format

Applications
• Strand displacement amplification of genomic DNA, plasmids, and phage DNA
• DNA labeling by random priming

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypePrimer
Shipping ConditionDry Ice
Concentration500 μM
PrimerRandom
Quantity100 μL
For Use With (Application)DNA Labeling, Strand Displacement Amplification
FormLiquid
Unit Size100 µL
Contents & Storage
20X Exo-Resistant Random Primer (100 μL)

Store at –20°C.