NoLimits 3500 bp DNA Fragment
Inquire about OEM or Commercial Supply version of this product here.
NoLimits 3500 bp DNA Fragment
Thermo Scientific™
NoLimits 3500 bp DNA Fragment
Green features
Thermo Scientific NoLimits is a collection of 36 highly-purified individual DNA fragments ranging from 10 to 20,000 bp. Unlike otherRead more
Have Questions?
Catalog number SM1501
Price (USD)/ 10 µg
85.60
Special Offer
Online exclusive
Ends: 16-Dec-2024
99.00 
Save 13.40 (14%)
-
Add to cart
Request bulk or custom format
Price (USD)/ 10 µg
85.60
Special Offer
Online exclusive
Ends: 16-Dec-2024
99.00 
Save 13.40 (14%)
Add to cart
NoLimits 3500 bp DNA Fragment
Catalog numberSM1501
Price (USD)/ 10 µg
85.60
Special Offer
Online exclusive
Ends: 16-Dec-2024
99.00 
Save 13.40 (14%)
-
Add to cart
Thermo Scientific NoLimits is a collection of 36 highly-purified individual DNA fragments ranging from 10 to 20,000 bp. Unlike other vendors, where custom standards are produced by PCR, Thermo Scientific NoLimits DNA fragments are produced using specifically designed plasmids purified by patented technology.Plasmid DNA, digested with Thermo Scientific Restriction Enzymes, is chromatography-purified to prepare the individual DNA fragments from the digestion mixture. The results are extremely pure DNA fragments, free of any truncated or degraded molecules. These custom DNA fragments are the best custom standards for applications ranging from electrophoresis (both gel and capillary) to HPLC and beyond.
Features
• Broad range: 10-20,000 bp
• Sharp peaks during capillary electrophoresis, sharp bands during gel electrophoresis
• Precise DNA concentration
• Chromatography-purified individual DNA fragments
Applications
• Gel electrophoresis
• Capillary electrophoresis
• DNA quantification
• HPLC analysis
• Applications where a custom DNA size standard is required.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel TypeAgarose
Product TypeDNA Fragment
Ready to LoadNo
Size Range10 to 20000 bp
Gel CompatibilityAgarose Gels
Green FeaturesSustainable packaging
Product LineNoLimits
Quantity10 μg
Unit Size10 µg