GeneRuler High Range DNA Ladder
These products are now shipping under ambient conditions. Learn more.
Inquire about OEM or Commercial Supply version of this product here.
GeneRuler High Range DNA Ladder
Thermo Scientific™
GeneRuler High Range DNA Ladder
Green features
Thermo Scientific GeneRuler High Range DNA Ladder is recommended for fast sizing and approximate quantification of high molecular weight double-strandedRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
SM13525 x 50 μg
SM135150 μg
Catalog number SM1352
Price (USD)/ 5 x 50 µg
351.48
Special Offer
Online exclusive
Ends: 16-Dec-2024
489.00 
Save 137.52 (28%)
-
Add to cart
Quantity:
5 x 50 μg
Request bulk or custom format
Price (USD)/ 5 x 50 µg
351.48
Special Offer
Online exclusive
Ends: 16-Dec-2024
489.00 
Save 137.52 (28%)
Add to cart
GeneRuler High Range DNA Ladder
Catalog numberSM1352
Price (USD)/ 5 x 50 µg
351.48
Special Offer
Online exclusive
Ends: 16-Dec-2024
489.00 
Save 137.52 (28%)
-
Add to cart
Thermo Scientific GeneRuler High Range DNA Ladder is recommended for fast sizing and approximate quantification of high molecular weight double-stranded DNA on agarose gels (in 1.5 hour, 0.4% agarose gel). The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler High Range DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase (see Protocol in the Resources.)
The ladder is supplied with 6X Orange DNA Loading Dye for sample DNA.
Highlights
• Ideal for both DNA sizing and approximate quantification
• Ideal for fast sizing of high molecular weight DNA fragments
• Sharp bands
• Supplied with loading dye for sample DNA
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration6X
Gel TypeAgarose
Label or DyeXylene Cyanol FF, Orange G
Product TypeHigh Range DNA Ladder
Ready to LoadNo
Size Range10171 to 48502 bp
Gel CompatibilityAgarose Gels
Green FeaturesSustainable packaging
Product LineGeneRuler
Quantity5 x 50 μg
Unit Size5 x 50 µg