MassRuler Express Forward DNA Ladder Mix, ready-to-use
These products are now shipping under ambient conditions. Learn more.
Inquire about OEM or Commercial Supply version of this product here.
MassRuler Express Forward DNA Ladder Mix, ready-to-use
Thermo Scientific™

MassRuler Express Forward DNA Ladder Mix, ready-to-use

Green features
Thermo Scientific MassRuler Express Forward and Reverse DNA Ladder mixes are specially designed for fast and reliable DNA quantification inRead more
Have Questions?
Catalog number SM1283
Price (USD)/ 2 x 500 µL
130.40
Special Offer
Online exclusive
Ends: 16-Dec-2024
163.00 
Save 32.60 (20%)
-
Add to cart
Request bulk or custom format
Price (USD)/ 2 x 500 µL
130.40
Special Offer
Online exclusive
Ends: 16-Dec-2024
163.00 
Save 32.60 (20%)
Add to cart
MassRuler Express Forward DNA Ladder Mix, ready-to-use
Catalog numberSM1283
Price (USD)/ 2 x 500 µL
130.40
Special Offer
Online exclusive
Ends: 16-Dec-2024
163.00 
Save 32.60 (20%)
-
Add to cart
Thermo Scientific MassRuler Express Forward and Reverse DNA Ladder mixes are specially designed for fast and reliable DNA quantification in short separation distances under various electrophoresis conditions (see Table 1). In the forward versions, the mass of each DNA fragment is directly proportional to the fragment size whereas, in the reverse versions, the mass is inversely proportional to fragment's size.

The intensity of the fragment in each ladder is calibrated against a standard that guarantees the precise quantity of each band. The ladders are mixtures of chromatography-purified, individual DNA fragments.

Highlights
• Accurate DNA sizing and quantification—DNA ladder fragments are quantified spectrophotometrically
• Fast and reliable DNA quantification — short separation distances under various electrophoresis conditions
• Sharp bands
• Easy-to-remember fragment sizes and quantities
• Ready-to-use—premixed with 6X MassRuler DNA Loading Dye for direct loading and room temperature storage
• Supplied with loading dye for sample DNA
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration6X
Gel TypeAgarose
Label or DyeBromophenol Blue
Product TypeDNA Ladder Mix
Ready to LoadYes
Shelf Life6 Months
Size Range100 to 1000 bp
Gel CompatibilityAgarose Gels
Green FeaturesSustainable packaging
Product LineMassRuler
Quantity2 x 500 μL
Unit Size2 x 500 µL