MassRuler Express LR Forward DNA Ladder, ready-to-use
These products are now shipping under ambient conditions. Learn more.
Inquire about OEM or Commercial Supply version of this product here.
MassRuler Express LR Forward DNA Ladder, ready-to-use
Thermo Scientific™

MassRuler Express LR Forward DNA Ladder, ready-to-use

Green features
Thermo Scientific MassRuler Express LR Forward and Reverse DNA ladders are specially designed for fast and reliable DNA quantification inRead more
Have Questions?
Catalog number SM1263
Price (USD)/ 2 x 500 µL
127.66
Special Offer
Online exclusive
Ends: 16-Dec-2024
138.00 
Save 10.34 (7%)
-
Add to cart
Request bulk or custom format
Price (USD)/ 2 x 500 µL
127.66
Special Offer
Online exclusive
Ends: 16-Dec-2024
138.00 
Save 10.34 (7%)
Add to cart
MassRuler Express LR Forward DNA Ladder, ready-to-use
Catalog numberSM1263
Price (USD)/ 2 x 500 µL
127.66
Special Offer
Online exclusive
Ends: 16-Dec-2024
138.00 
Save 10.34 (7%)
-
Add to cart
Thermo Scientific MassRuler Express LR Forward and Reverse DNA ladders are specially designed for fast and reliable DNA quantification in short separation distances under various electrophoresis conditions. In the forward versions, the mass of each DNA fragment is directly proportional to the fragment size, whereas in the reverse versions, the mass is inversely proportional to fragment's size.

The intensity of the fragment in each ladder is calibrated against a standard that guarantees the precise quantity of each band. The ladders are mixtures of chromatography-purified, individual DNA fragments.

Highlights

• Accurate DNA sizing and quantification—DNA ladder fragments are quantified spectrophotometrically
• Fast and reliable DNA quantification—short separation distances under various electrophoresis conditions
• Sharp bands
• Easy-to-remember fragment sizes and quantities
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration6X
Gel TypeAgarose
Label or DyeBromophenol Blue
Product TypeDNA Ladder
Ready to LoadYes
Shelf Life6 Months
Size Range100 to 1000 bp
Gel CompatibilityAgarose Gels
Green FeaturesSustainable packaging
Product LineMassRuler
Quantity2 x 500 μL
Unit Size2 x 500 µL