O'RangeRuler 100 bp DNA Ladder, ready-to-use
These products are now shipping under ambient conditions. Learn more.
Inquire about OEM or Commercial Supply version of this product here.
O'RangeRuler 100 bp DNA Ladder, ready-to-use
Thermo Scientific™

O'RangeRuler 100 bp DNA Ladder, ready-to-use

Green features
Thermo Scientific O'RangeRuler 100 bp DNA Ladder 100-1500 bp is designed for precise sizing of PCR products and other double-strandedRead more
Have Questions?
Catalog number SM0623
Price (USD)/ 25 µg
83.65
Online exclusive
109.00 
Save 25.35 (23%)
-
Add to cart
Request bulk or custom format
Price (USD)/ 25 µg
83.65
Online exclusive
109.00 
Save 25.35 (23%)
Add to cart
O'RangeRuler 100 bp DNA Ladder, ready-to-use
Catalog numberSM0623
Price (USD)/ 25 µg
83.65
Online exclusive
109.00 
Save 25.35 (23%)
-
Add to cart
Thermo Scientific O'RangeRuler 100 bp DNA Ladder 100-1500 bp is designed for precise sizing of PCR products and other double-stranded DNA fragments in agarose or non-denaturing polyacrylamide gels. It is step ladder with 100 bp differences between the DNA fragments. It consists of purified and ligated blunt ended basic unit repeats of 100 bp. Not recommended for DNA quantification.

O'RangeRuler 100 bp DNA Ladder is supplied with 6X Orange DNA Loading Dye for sample DNA.

Highlights

• Step ladder with 100 bp increments
• Sharp bands, easy-to-remember fragment sizes
• Evenly spaced reference bands
• Ready-to-use—premixed with 6X Orange Loading Dye for direct loading and room temperature storage
• Supplied with loading dye for sample DNA
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration6X
Gel TypeAgarose
Label or DyeXylene Cyanol FF, Orange G
Product TypeDNA Ladder
Ready to LoadYes
Shelf Life6 Months
Size Range100 to 500 bp
Gel CompatibilityAgarose Gels
Green FeaturesSustainable packaging
Product LineO’GeneRuler, O’RangeRuler
Quantity25 μg
Unit Size25 µg