Thermo Scientific GeneRuler 50 bp DNA Ladder is recommended for sizing and approximate quantification of a double-stranded DNA in the range of 50 bp to 1,000 bp on agarose or polyacrylamide gels. The DNA ladder consists of 13 DNA fragments and is provided with 6X TriTrack DNA Loading Dye.
Highlights of 50 bp DNA Ladder
• Easy visualization—three-color tracking of DNA migration during electrophoresis with 6X TriTrack DNA Loading Dye (contains Xylene Cyanol FF, Bromophenol Blue, and Orange G)
• Simplicity—one loading dye for a variety of samples
• Sharp bands—clear results with mixtures of individual chromatography-purified DNA fragments
Product use
This double-stranded DNA ladder is compatible with 1–2% standard and precast agarose slab gels and can be visualized after ethidium bromide or SYBR Safe staining. A uniform intensity of bands and bright reference bands allow easy sizing and orientation. A defined amount of DNA in each band enables approximate quantification of sample DNA.
For Research Use Only. Not for use in diagnostic procedures.