Specifications
Concentration6X
Gel TypeAgarose
Label or DyeXylene Cyanol FF, Bromophenol Blue, Orange G
Product TypeDNA Ladder Mix
Ready to LoadYes
Shelf Life6 Months
Size Range100 to 1000 bp
Gel CompatibilityAgarose Gels
Green FeaturesSustainable packaging
Product LineGeneRuler
Quantity100 Applications
Unit Size50 µg