Thermolyne™ Premium Large Muffle Furnaces Accessories
Product Image
Thermo Scientific™

Thermolyne™ Premium Large Muffle Furnaces Accessories

Use these accessories with Thermolyne Premium Large Muffle Furnaces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
SH408X1Stainless-steel Shelf
JSX16Shelf Pegs
PH177X1Hearth Tray, 22.9 x 27.3 x 1.9 cm
PHX1Hearth Tray, 8.2 x 10.1 x 1.27 cm
Catalog number SH408X1
Price (USD)/ Each
-
Request A Quote
Description:
Stainless-steel Shelf

Use Thermo Scientific™ Thermolyne Premium Large Muffle Furnace Accessories to increase the utility of the Large Muffle Furnace.

Specifications
DescriptionStainless-steel Shelf
For Use With (Equipment)Thermolyne Premium Large Muffle Furnace
TypeShelf
Length (English)8.3 in.
Width (English)12.8 in.
Unit SizeEach