Sodium Green™, Tetra (Tetramethylammonium) Salt, cell impermeant
Sodium Green™, Tetra (Tetramethylammonium) Salt, cell impermeant
Invitrogen™

Sodium Green™, Tetra (Tetramethylammonium) Salt, cell impermeant

This cell-impermeant form of Sodium Green™, tetra(tetramethylammonium) salt is a visible light-excitable Na+ indicator.Learn more about ion indicators including calcium,Read more
Have Questions?
Catalog number S6900
also known as S-6900
Price (USD)/ 1 mg
395.00
-
Add to cart
Price (USD)/ 1 mg
395.00
Add to cart
Sodium Green™, Tetra (Tetramethylammonium) Salt, cell impermeant
Catalog numberS6900
Price (USD)/ 1 mg
395.00
-
Add to cart
This cell-impermeant form of Sodium Green™, tetra(tetramethylammonium) salt is a visible light-excitable Na+ indicator.

Learn more about ion indicators including calcium, potassium, pH, and membrane potential indicators ›

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer
Product TypeSodium Indicator
Dye TypeSodium Indicator
Product LineSodium Green™
Quantity1 mg
Shipping ConditionRoom Temperature
Unit Size1 mg
Contents & Storage
Store at room temperature and protect from light.