Sulforhodamine 101 is a water-soluble, nonfixable red fluorescent (excitation/emission maxima ∼ 586/605 nm) polar tracer that can be used forRead more
Have Questions?
Catalog number S359
Price (USD)/ 25 mg
156.65
Online exclusive
160.00
Save 3.35 (2%)
-
Add to cart
Price (USD)/ 25 mg
156.65
Online exclusive
160.00
Save 3.35 (2%)
Add to cart
Sulforhodamine 101
Catalog numberS359
Price (USD)/ 25 mg
156.65
Online exclusive
160.00
Save 3.35 (2%)
-
Add to cart
Sulforhodamine 101 is a water-soluble, nonfixable red fluorescent (excitation/emission maxima ∼ 586/605 nm) polar tracer that can be used for investigating neuronal morphology, preparing fluorescent liposomes, studying cell-cell communications and following phagosome-lysosome uptake and fusion.
For Research Use Only. Not for use in diagnostic procedures.