QuEChERS EN 15662 Method Clean-up Kits
QuEChERS EN 15662 Method Clean-up Kits
Actual product may vary
Thermo Scientific™

QuEChERS EN 15662 Method Clean-up Kits

Use Thermo Scientific™ QuEChERS EN 15662 Method Clean-up Kits to analyze complex sample matrices for residues, toxins, and other contaminants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionSalt CompositionSample TypeTube Size
S2-2-FW-EN-KITQuEChERS EN 15662 Method Clean-up Kit; 150 mg MgSO4, 25 mg PSA, 25 mg C18 prefilled in 2 mL Tubes; 50/Pk.150 mg MgSO4, 25 mg PSA, 25 mg C18Fruits and Vegetables with Fats and Waxes2 mL
S2-15-FW-EN-KITQuEChERS EN 15662 Method Clean-up Kit; 900 mg MgSO4, 150 mg PSA, 150 mg C18 prefilled in 15 mL Tubes; 50/Pk.900 mg MgSO4, 150 mg PSA, 150 mg C18Fruits and Vegetables with Fats and Waxes15 mL
S2-15-HP-EN-KITQuEChERS EN 15662 Method Clean-up Kit; 900 mg MgSO4, 150 mg PSA, 45 mg GCB prefilled in 15 mL Tubes; 50/Pk.900 mg MgSO4, 150 mg PSA, 45 mg GCBHighly Pigmented Fruits and Vegetables15 mL
S2-15-P-EN-KITQuEChERS EN 15662 Method Clean-up Kit; 900 mg MgSO4, 150 mg PSA, 15 mg GCB prefilled in 15 mL Tubes; 50/Pk.900 mg MgSO4, 150 mg PSA, 15 mg GCBPigmented Fruits and Vegetables15 mL
S2-2-GFV-EN-KITQuEChERS EN 15662 Method Clean-up Kit; 150 mg MgSO4, 25 mg PSA prefilled in 2 mL Tubes; 100/Pk.150 mg MgSO4, 25 mg PSAGeneral Fruits and Vegetables2 mL
S2-2-HP-EN-KITQuEChERS EN 15662 Method Clean-up Kit; 150 mg MgSO4, 25 mg PSA, 7.5 mg GCB prefilled in 2 mL Tubes; 100/Pk.150 mg MgSO4, 25 mg PSA, 7.5 mg GCBHighly Pigmented Fruits and Vegetables2 mL
S2-2-P-EN-KITQuEChERS EN 15662 Method Clean-up Kit; 150 mg MgSO4, 25 mg PSA, 2.5 mg GCB prefilled in 2 mL Tubes; 100/Pk.150 mg MgSO4, 25 mg PSA, 2.5 mg GCBPigmented Fruits and Vegetables2 mL
Catalog number S2-2-FW-EN-KIT
Price (USD)/ Pack of 100
183.46
-
Add to cart
Description:
QuEChERS EN 15662 Method Clean-up Kit; 150 mg MgSO4, 25 mg PSA, 25 mg C18 prefilled in 2 mL Tubes; 50/Pk.
Salt Composition:
150 mg MgSO4, 25 mg PSA, 25 mg C18
Sample Type:
Fruits and Vegetables with Fats and Waxes
Tube Size:
2 mL
Price (USD)/ Pack of 100
183.46
Add to cart
QuEChERS EN 15662 Method Clean-up Kits
Catalog numberS2-2-FW-EN-KIT
Price (USD)/ Pack of 100
183.46
-
Add to cart
Use Thermo Scientific™ QuEChERS Clean-up Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis of pesticide residues, veterinary drug residues, and toxins using the EN 15662 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.

Thermo Scientific™ QuEChERS EN 15662 Method Clean-Up Kits make sample preparation straight-forward and efficient, enabling you to confidently perform extractions and analyze fruit and vegetable samples for pesticide residues and other contaminants.

Clean-up kits are available in prefilled 2 mL and 15 mL tube formats for the following matrices: general fruits and vegetables, pigmented fruits and vegetables, highly pigmented fruits and vegetables, and fruits and vegetables containing fats and waxes.

What is the purpose of the chemicals used in the EN 15662 Method?

  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer; removes remaining water from sample
  • NaCl: induces phase separation between water content in sample and acetonitrile layer; removes residual water from sample
  • Trisodium Citrate: buffers the sample to stabilize pH
  • Disodium Citrate Sesquihydrate: buffers the sample to stabilize pH
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids, from sample
  • C18: removes fats, sterols, and other non-polar interferences from sample
  • GCB: (graphitized carbon black) removes pigment (not recommended for use with planar pesticides)
Specifications
DescriptionQuEChERS EN 15662 Method Clean-up Kit; 150 mg MgSO4, 25 mg PSA, 25 mg C18 prefilled in 2 mL Tubes; 50/Pk.
TypeQuEChERS Clean-up Kit
IncludesSalts in Prefilled Tubes
MethodQuEChERS sample preparation for EN 15662 Method
Salt Composition150 mg MgSO4, 25 mg PSA, 25 mg C18
Sample TypeFruits and Vegetables with Fats and Waxes
Tube Size2 mL
Unit SizePack of 100