NY/T Method QuEChERS Clean-up Kit for 15 g Samples, General Fruits and Vegetables
NY/T Method QuEChERS Clean-up Kit for 15 g Samples, General Fruits and Vegetables
Thermo Scientific™
NY/T Method QuEChERS Clean-up Kit for 15 g Samples, General Fruits and Vegetables
Use Thermo Scientific™ QuEChERS NY/T 1380-2007 Method Clean-up Kits for accurate determination of 51 specific pesticide residues in fruits and vegetables.
Have Questions?
Catalog number S2-15-GFV-NYT-KIT
Price (USD)/ Pack of 50
157.47
-
Add to cart
Description:
QuEChERS NY/T 1380-2007 Method Clean-up Kit; 100 mg PSA, 100 mg C18, 300 mg Anhydrous MgSO4 prefilled in 15 mL Tubes; 50/Pk.
Salt Composition:
300 mg Anhydrous MgSO4, 100 mg PSA, 100 mg C18
Sample Type:
General Fruits and Vegetables
Tube Size:
15 mL
Price (USD)/ Pack of 50
157.47
Add to cart
NY/T Method QuEChERS Clean-up Kit for 15 g Samples, General Fruits and Vegetables
Catalog numberS2-15-GFV-NYT-KIT
Price (USD)/ Pack of 50
157.47
-
Add to cart
Use Thermo Scientific™ QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) NY/T 1380-2007 Method Clean-up Kits for accurate and efficient determination of 51 specific pesticide residues in fruits and vegetables. Our QuEChERS NY/T 1380-2007 Method Clean-up Kits are ideal for preparing samples prior to GC-MS and can be automated for higher throughput.
The NY/T 1380-2007 Method supports voluntary testing of professional and industrial standards for agriculture in China. Thermo Scientific™ QuEChERS NY/T 1380-2007 Method Clean-up Kits make sample preparation straight-forward and efficient, allowing for determination of pesticide residues in fruit and vegetable matrices.
We offer a clean-up/dispersive SPE kit composed of pre-filled 15 mL tubes for use with general fruit and vegetable matrices.
What is the purpose of the chemicals used in the NY/T 1380-2007 Method?
  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer during liquid-liquid partitioning step; removes remaining water from sample during dispersive solid phase extraction (dSPE) step
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids, from sample during dispersive solid phase extraction (dSPE) step
  • C18: removes fats, sterols, and other non-polar interferences from sample
Specifications
DescriptionQuEChERS NY/T 1380-2007 Method Clean-up Kit; 100 mg PSA, 100 mg C18, 300 mg Anhydrous MgSO4 prefilled in 15 mL Tubes; 50/Pk.
TypeQuEChERS Clean-up Kit
IncludesSalts in Prefilled Tubes
MethodQuEChERS sample preparation for NY/T 1380-2007 Method
Salt Composition300 mg Anhydrous MgSO4, 100 mg PSA, 100 mg C18
Sample Size15 g sample
Sample TypeGeneral Fruits and Vegetables
Tube Size15 mL
Unit SizePack of 50