QuEChERS AOAC 2007.01 Method Extraction Kits
QuEChERS AOAC 2007.01 Method Extraction Kits
Actual product may vary
Thermo Scientific™

QuEChERS AOAC 2007.01 Method Extraction Kits

Use Thermo Scientific™ QuEChERS AOAC 2007.01 Method Extraction Kits to prepare complex samples for LC-MS or GC-MS analysis of residues, toxins, and other contaminants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionIncludesSalt CompositionTube Size
S1-15-AOAC-POTQuEChERS AOAC Extraction Kit for 15 g sample; 6 g MgSO4, 1.5 g NaOAC in Pouches; No Tubes; 50/Pk.Salts in Pouches6 g MgSO4, 1.5 g NaOACNone
S1-15-AOAC-CH-KITQuEChERS AOAC Extraction Kit for 15 g sample; 6 g MgSO4, 1.5 g NaOAc in Pouches; Ceramic Homogenizers; Empty 50 mL Tubes; 50/Pk.Salts in Pouches, Tubes, Ceramic Homogenizers6 g MgSO4, 1.5 g NaOAc50 mL
S1-15-AOAC-KITQuEChERS AOAC Extraction Kit for 15 g sample; 6 g Anhydrous MgSO4, 1.5 g NaOAc in Pouches; Empty 50 mL Tubes; 50/Pk.Salts in Pouches, Tubes6 g Anhydrous MgSO4, 1.5 g NaOAc50 mL
Catalog number S1-15-AOAC-POT
Price (USD)/ Pack of 50
168.71
-
Add to cart
Description:
QuEChERS AOAC Extraction Kit for 15 g sample; 6 g MgSO4, 1.5 g NaOAC in Pouches; No Tubes; 50/Pk.
Includes:
Salts in Pouches
Salt Composition:
6 g MgSO4, 1.5 g NaOAC
Tube Size:
None
Price (USD)/ Pack of 50
168.71
Add to cart
QuEChERS AOAC 2007.01 Method Extraction Kits
Catalog numberS1-15-AOAC-POT
Price (USD)/ Pack of 50
168.71
-
Add to cart
Use Thermo Scientific™ QuEChERS Extraction Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis of pesticide residues, veterinary drug residues, and toxins using the AOAC 2007.01 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.

QuEChERS AOAC 2007.01 Method Extraction Kits

Extract analytes of interest from a homogenized sample using extraction salts. Use these kits with 10 g or 15 g samples. Choose from:

  • Extraction Salts in Pouches, No Tubes
  • Extraction Salts in Pouches with 50 mL Tubes
  • Extraction Salts in Pouches with 50 mL Tubes and Ceramic Homogenizers

Chemicals used in the AOAC 2007.01 Method

  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer during liquid-liquid partitioning; removes remaining water from sample during dSPE
  • NaOAc: buffers the sample to stabilize pH
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids from sample during dispersive solid phase extraction
  • C18: removes fats, sterols, and other non-polar interferences from sample
  • GCB: graphitized carbon black, removes pigment (not recommended for use with planar pesticides)
Specifications
DescriptionQuEChERS AOAC Extraction Kit for 15 g sample; 6 g MgSO4, 1.5 g NaOAC in Pouches; No Tubes; 50/Pk.
TypeQuEChERS Extraction Kit
IncludesSalts in Pouches
MethodQuEChERS sample preparation for AOAC 2007.01 Method
Salt Composition6 g MgSO4, 1.5 g NaOAC
Sample Size15 g sample
Tube SizeNone
Unit SizePack of 50