Savant™ SpeedVac™ Refrigerated Vapor Trap Series
Product Image
Thermo Scientific™
Savant™ SpeedVac™ Refrigerated Vapor Trap Series
Condense and trap a wide variety of solvent vapors that can contaminate and degrade vacuum pump oil with Thermo Scientific™ Savant™ Refrigerated Vapor Traps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionElectrical Requirements
RVT450-230Refrigerated Vapor Cold Trap RVT450230 V 50 Hz
RVT450-115Refrigerated Vapor Cold Trap RVT450115 V 60 Hz
Catalog number RVT450-230
Price (USD)/ Each
-
Request A Quote
Description:
Refrigerated Vapor Cold Trap RVT450
Electrical Requirements:
230 V 50 Hz
Condenses and removes a wide variety of solvent vapors that can contaminate and degrade vacuum pump oil with Thermo Scientific™ Savant™ Refrigerated Vapor Traps (RVT). The RVT safely recovers the solvent and reduces emissions of hazardous chemicals in the environment.
Solvents are removed from samples during vacuum evaporation with a SpeedVac™ Concentrator which are then collected in a CFC-free Refrigerated Vapor Trap (RVT).   Solvent vapors pass into a Glass Condensation Flask and condense on its walls. This prevents the vapors from reaching and potentially damaging the oil-sealed vacuum pump. 
  • Rapid cooling time minimizes waiting period
  • Easy to remove glass flask for quick solvent changes
  • Rapidly cools to operating temperature
  • Easy to use glass condensation flask allows for the collection of liquid into a self-contained jar for easy draining
  • Sealed refrigeration system eliminates consumable coolants such as dry ice and acetone
Specifications
Certifications/ComplianceUL Listed to US and Canadian requirements. CE marked
DescriptionRefrigerated Vapor Cold Trap RVT450
TypeRefrigerated Vapor Trap
Electrical Requirements230 V 50 Hz
Unit SizeEach