RapID™ ANA II System
RapID™ ANA II System
Thermo Scientific™
RapID™ ANA II System
Identify over 90 medically important anaerobes in just four hours using this rapid identifcation system based on enzyme technology.
Have Questions?
Catalog number R8311002
Price (USD)/ Each
-
Request A Quote
Description:
RapID ANA II System
Detectable Analytes:
Anaerobes
Quantity:
20 Panels/Kit
Identify over 90 medically important anaerobes in just four hours using the Thermo Scientific™ RapID™ ANA II System, an identification system based on enzyme technology.
The RapID ANA II System is a qualitative micromethod employing conventional and chromogenic substrates for the identification of medically important anaerobic bacteria isolated from human clinical specimens.
  • One-step inoculation—decreased prep time for increased productivity.
  • Manual microbial identification made easy—no oil, no pipetting, no 24-hr incubation
  • Four-hour incubation*—quicker time-to-result for a faster response
  • Common procedure—lessened materials usage saves time and money
  • Visible color reactions—reduced subjectivity decreases repeat testing
  • Better reporting—more coverage, advanced updates
*Two-hour incubation for RapID SS/u.
A clear plastic tray contains reagent impregnated wells, allowing simultaneous inoculation of each cavity with a predetermined volume of inoculum. A suspension of test organism in RapID Inoculation Fluid is used as the inoculum which rehydrates and initiates test reactions.
After incubation, each cavity is examined for reactivity by noting the development of a color. In some cases, reagents must be added to the cavities to provide a color change. The resulting pattern of positive and negative scores is used as the basis for identification of the test isolate by comparison of test results to reactivity patterns stored in a database, or through the use of a computer-generated Code Compendium (ERIC).
Requires:
RapID Spot Indole, Part No. R8309002, and RapID Inoculation Fluid, Part No. R8325102
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionRapID ANA II System
CE MarkerYes
Detectable AnalytesAnaerobes
Packaging Type1 Box
Quantity20 Panels/Kit
Unit SizeEach
Contents & Storage
2°C to 8°C