Quanti-Cult™ Pseudomonas paraeruginosa ATCC™ 9027™
Quanti-Cult™ Pseudomonas paraeruginosa ATCC™ 9027™
Thermo Scientific™
Quanti-Cult™ Pseudomonas paraeruginosa ATCC™ 9027™
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Have Questions?
Catalog number R4735210
Price (USD)/ Each
-
Request A Quote
Description:
Pseudomonas paraeruginosa ATCC™ 9027™
Thermo Scientific™ Quanti-Cult™ Pseudomonas paraeruginosa ATCC™ 9027™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial, ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.3mL inoculum
Ordering Alerts:
Yield is 1 Test/reconstituted vial, 10 Tests/kit
Includes:
10 preserved organism vials and 10 rehydration vials.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®9027TM*
DescriptionPseudomonas paraeruginosa ATCC™ 9027™
FormatQuanti-Cult
Product TypeBactidisk
Target Organism ClassGram-negative Rods
Test TypeQuantitative QC
Packaging Type10 Test/Kit
Unit SizeEach