Quanti-Cult Plus™ Burkholderia multivorans ATCC™ BAA-247™
Quanti-Cult Plus™ Burkholderia multivorans ATCC™ BAA-247™
Thermo Scientific™

Quanti-Cult Plus™ Burkholderia multivorans ATCC™ BAA-247™

Quality control to comply with USP chapter 60
Have Questions?
Catalog number R4715222
Price (USD)/ Pack of 100
-
Request A Quote
Thermo Scientific™ Quanti-Cult Plus™ Burkholderia multivorans ATCC™ BAA-247™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures. It is the recommended strain to support QC testing for non-sterile products described in US Pharmacopeia chapter 60.
No serial dilution required.
 
  • Delivers <100 colony forming units (CFUs) in a single 0.1mL inoculum
  • 10 tests/reconstituted vial; 100 tests/kit
  • Ready for rehydration
  • For use in quality control procedures

Includes:
10 preserved organism vials and 10 rehydration vials. 
 
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
For Laboratory Use
Specifications
ATCC®ATCC™ BAA-247™
DescriptionBurkholderia multivorans ATCC™ BAA-247™
FormatQuanti-Cult Plus™
Product TypeBactidisk
Target Organism ClassGram-negative Rods
Test TypeQuantitative QC
For Use With (Application)Pharmaceutical
Packaging Type100 Test/Kit
Quantity100 Tests/Kit
Strain DesignationsLMG 13010, CCUG 34080, CIP 105495, DSM 13243, NCTC 13007
Unit SizePack of 100