Culti-Loops™ Proteus vulgaris ATCC™ 8427™
Culti-Loops™ Proteus vulgaris ATCC™ 8427™
Thermo Scientific™
Culti-Loops™ Proteus vulgaris ATCC™ 8427™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Have Questions?
Catalog number R4607058
Price (USD)/ Each
-
Request A Quote
Quantity:
5 Loops/Pack
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®8427TM*
DescriptionProteus vulgaris ATCC™ 8427™
FormatCulti-Loops
Product TypeCulti-Loops Quality Control Organism
Target Organism ClassGram-negative Rods
Test TypeQualitative QC
Quantity5 Loops/Pack
Unit SizeEach