You need to enable JavaScript to run this app.
Remel™ Egg Yolk Tellurite 100%
Actual product may vary
Thermo Scientific™
Remel™ Egg Yolk Tellurite 100%
Remel Egg Yolk Tellurite 100% is used with Baird Parker Agar Base.
Have Questions?
Change view
Catalog Number
Quantity
Packaging Type
R450330
100 mL
1 Ea.
R450331
100 mL
10/Pk.
2 Options
Catalog number
R450330
Price (USD)
/ Each
-
Request A Quote
Quantity:
100 mL
Packaging Type:
1 Ea.
Remel Egg Yolk Tellurite 100% is used with Baird Parker Agar Base.
Specifications
Description
Egg Yolk Tellurite 100%
Form
Liquid
Product Type
Microbiology Media
Packaging Type
1 Ea.
Quantity
100 mL
Unit Size
Each