Contents & Storage
Provided as 1-mL solution at 30 x concentration in a LDPE dropper bottle.
Store (at 2°C to 8°C
When stored as directed, product can be used for 6 months after receipt
.One bottle is sufficient to treat approximately 125-250 tissue sections.
)