SYTOX AADvanced Ready Flow Reagent is a high-affinity nucleic acid stain for the detection of dead cells in flow cytometry.Read more
Have Questions?
Catalog number R37173
Price (USD)/ 120 reactions
203.65
Online exclusive
215.00
Save 11.35 (5%)
-
Add to cart
Price (USD)/ 120 reactions
203.65
Online exclusive
215.00
Save 11.35 (5%)
Add to cart
SYTOX AADvanced™ Ready Flow™ Reagent
Catalog numberR37173
Price (USD)/ 120 reactions
203.65
Online exclusive
215.00
Save 11.35 (5%)
-
Add to cart
SYTOX AADvanced Ready Flow Reagent is a high-affinity nucleic acid stain for the detection of dead cells in flow cytometry. The dye has excitation/emission maxima of 546/647 nm.
• Stable at room temperature • Convenient, ready-to-use format—no need to dilute, weigh, or pipette • Rapid uptake and low coefficient of variation (CV)
SYTOX AADvanced Ready Flow Reagent is a ready-to-use, direct-to-sample dead cell stain that emits red fluorescence when bound to DNA. Add 2 drops form the dropper bottle directly into 1 x 106 cells, incubate, and analyze.
The SYTOX AADvanced Ready Flow Reagent dye is spectrally similar to 7-AAD, but with rapid uptake kinetics and relatively low CVs . It penetrates cells more efficiently than 7-AAD, providing better separation of live and dead cells.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityImpermeant
ColorRed
DescriptionSYTOX AADvanced™ Ready Flow™ Reagent
Detection MethodFluorescence
For Use With (Application)Viability Assay
For Use With (Equipment)Flow Cytometer
Product TypeReagent
Dye TypeCell-Impermeant
Emission647 nm
Excitation Wavelength Range546 nm
FormSolution
FormatBottle(s)
Product LineAADvanced™, Ready Flow™
Quantity120 Reactions
Shipping ConditionRoom Temperature
SolubilityDMSO (Dimethylsulfoxide)
Sub Cellular LocalizationNucleus
Unit Size120 reactions
Contents & Storage
Includes 2 dropper bottles, store at room temperature.