SYTOX™ Green Ready Flow™ Reagent
SYTOX™ Green Ready Flow™ Reagent
Invitrogen™

SYTOX™ Green Ready Flow™ Reagent

SYTOX™ Green Ready Flow™ Reagent is a bright, easy-to-use nucleic acid stain for distinguishing dead cells from live cells inRead more
Have Questions?
Catalog number R37168
Price (USD)/ 120 assays
293.65
Online exclusive
314.00 
Save 20.35 (6%)
-
Add to cart
Price (USD)/ 120 assays
293.65
Online exclusive
314.00 
Save 20.35 (6%)
Add to cart
SYTOX™ Green Ready Flow™ Reagent
Catalog numberR37168
Price (USD)/ 120 assays
293.65
Online exclusive
314.00 
Save 20.35 (6%)
-
Add to cart
SYTOX™ Green Ready Flow™ Reagent is a bright, easy-to-use nucleic acid stain for distinguishing dead cells from live cells in flow cytometry assays. It is excitable at 488 nm with an emission profile similar to FITC, and has been tested in multiple cell lines.

• Stable at room temperature
• Convenient, ready-to-use format—no need to dilute, weigh, or pipette
• Exceptionally bright green fluorescence upon binding to nucleic acids
• Rapid staining without wash steps

SYTOX Green Ready Flow Reagent is a ready-to-use, direct-to-sample dead cell stain. Add 2 drops from the dropper bottle directly into 1 x 106 cells, incubate, and analyze.

As a cell-impermeant dye, SYTOX Green enters the cell upon loss of membrane integrity and binds to DNA, thereby acting as a counterstain that can be analyzed when excited at 488 nm and the emission captured at a peak of 523 nm. SYTOX Green reagent can be used in both mammalian and bacterial cells and is often included in multiplex assays.

Specifications
Cell PermeabilityImpermeant
ColorGreen
DescriptionSYTOX™ Green Ready Flow™ Reagent
Detection MethodFluorescence
For Use With (Application)Viability Assay
For Use With (Equipment)Flow Cytometer
Product TypeReagent
Dye TypeSYTOX™ Green Ready Flow™
Emission523 nm
Excitation Wavelength Range504 nm
FormSolution
FormatBottle(s)
Product LineReady Flow™, Sytox™
Quantity120 Assays
SolubilityDMSO (Dimethylsulfoxide)
Unit Size120 assays
Contents & Storage
Includes 2 dropper bottles, store at room temperature.