PrimeStore Molecular Transport Medium
PrimeStore Molecular Transport Medium

PrimeStore Molecular Transport Medium

Non-hazardous, inactivation media for viral collection and transport with Inactivation media for nucleic acid testing and molecular diagnostics.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFill VolumeTube Size
R13902Primestore MTM Collection Tube (1mL), 50/bx1 mL2 mL
R13903PrimeStore MTM Collection Tube (1.5mL), 50/bx1.5 mL3 mL
R13905PrimeStore MTM Collection Tube (1.5mL), 50/bx1.5 mL5 mL
Catalog number R13902
Price (USD)/ Each
-
Request A Quote
Description:
Primestore MTM Collection Tube (1mL), 50/bx
Fill Volume:
1 mL
Tube Size:
2 mL

Stabilize, inactivate and transport infectious unprocessed nasal washes, nasopharyngeal, oral / throat swabs suspected of containing influenza A virus RNA with Longhorn PrimeStore Molecular Transport Medium™ (PS-MTM).

PrimeStore Molecular Transport Medium™ is also intended for the stabilization, transportation, and inactivation of infectious unprocessed sputum samples suspected of containing Mycobacterium tuberculosis (MTB) DNA from human samples.

Specimen collection and transport is a key component in the molecular detection of influenza A and MTB. PS-MTM is a self-contained, ‘ready-to-use’ system that allows for the stabilization and safe transport of clinical samples at ambient temperature from the collection site to the laboratory.

Distributed globally by Thermo Fisher Scientific
Specifications
Closure ColorClear
CompatibilityThe PS-MTM system is a collection, preservation, transport, and storage system for influenza RNA and MTB DNA. Extraction and purification of nucleic acids have been validated on several manual spin column kits (PrimeXtract™, RNAqueous Micro Kit, Viral RNA Mini Kit, QiaAMP DNA Mini Kit) and automated magnetic bead extraction kits (NucliSENS EasyMAG and MagNA Pure 96 System using the DNA Bacterial/Viral small volume kit). The user is responsible for validating additional extraction and purification kits and platforms.
DescriptionPrimestore MTM Collection Tube (1mL), 50/bx
Fill Volume1 mL
For Use With (Application)Intended for stabilization, transportation, and inactivation of infectious unprocessed nasal washes, nasopharyngeal, oral/throat swabs suspected of containing influenza A virus RNA. PS-MTM is also intended for the stabilization, transportation and inactivation of infectious unprocessed sputum samples suspected of containing Mycobacterium tuberculosis (MTB) DNA from human samples.
Quantity50/Pk.
Shelf Life24 Months
Storage RequirementsRoom temperature, 2°C to 25°C
Tube Size2 mL
Unit SizeEach