Remel™ Tryptic Soy Agar
Remel™ Tryptic Soy Agar
Actual product may vary
Thermo Scientific™

Remel™ Tryptic Soy Agar

The Thermo Scientific™ Remel Tryptic Soy Agar (TSA) (Soybean Casein Digest Agar) is a general purpose medium for cultivation and isolation of microorganisms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatPackaging Type
R11270010 x 500 mLBB - 500mL10/Pk.
R01917100/Pk.Monoplate100/Pk.
R0192010/Pk.Monoplate10/Pk.
R0408010/Pk.15mm x 150mm Plate10/Pk.
R064860100/Pk.15mm x 103mm Tube100/Pk.
R06486220/Pk.15mm x 103mm Tube20/Pk.
R08930100/Pk.20mm x 113mm Tube100/Pk.
R0893220/Pk.20mm x 113mm Tube20/Pk.
R08934100 x 10 mL20mm x 113mm Pour Tube100/Pk.
R09938100 x 22 mL20mm x 150mm Pour Tube100/Pk.
R0994020 x 22 mL20mm x 150mm Pour Tube20/Pk.
R110092100/Pk.15mm x 100mm Plate100/Pk.
R11270310 x 100 mLBRB - 100mL10/Pk.
R0192210/Pk.Monoplate Deep Pour10/Pk.
Catalog number R112700
Price (USD)/ Each
-
Request A Quote
Quantity:
10 x 500 mL
Format:
BB - 500mL
Packaging Type:
10/Pk.

Thermo Scientific™ Remel Tryptic Soy Agar (TSA) (Soybean Casein Digest Agar) is a general purpose medium for cultivation and isolation of microorganisms.

REF R110092, REF R112700, and REF R112703 not intended for IVD use.
Specifications
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
FormatBB - 500mL
Product TypeAgar
Packaging Type10/Pk.
Quantity10 x 500 mL
Unit SizeEach