Fluid Thioglycollate Medium with Indicator, Dextrose Medium
Fluid Thioglycollate Medium with Indicator, Dextrose Medium
Thermo Scientific™
Fluid Thioglycollate Medium with Indicator, Dextrose Medium
Reduce prep time while isolating aerobes, anaerobes and microaerophilic organisms with this liquid medium with formats available that meet international industry guidelines.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatMedia TypeManufacturing Origin
R11264610/Pk.BottleFluid Thioglycollate MediumUnited States
R064690100/Pk.TubeFluid Thioglycollate MediumUnited States
R07172100/Pk.16mm x 125mm TubeFluid Thioglycollate MediumUnited States
R08846100/Pk.20mm x 113mm TubeFluid Thioglycollate MediumUnited States
R11264010/Pk.WMB LugFluid Thioglycollate MediumUnited States
R11264110/Pk.SBFluid Thioglycollate MediumUnited States
R11264210/Pk.Borosilicate Bottle - SeptumFluid Thioglycollate MediumUnited States
R11780020/Pk.16mm x 125mm Tube, Hungate CapFluid Thioglycollate MediumUnited States
R117801100/Pk.16mm x 125mm Tube, Hungate CapFluid Thioglycollate MediumUnited States
Catalog number R112646
Price (USD)/ Each
-
Request A Quote
Quantity:
10/Pk.
Format:
Bottle
Media Type:
Fluid Thioglycollate Medium
Manufacturing Origin:
United States
Cultivate and isolate aerobes, anaerobes and microaerophilic organisms using Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium.
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes and bottles of Fluid Thioglycollate Medium w/Indicator, Dextrose.
Fluid Thioglycollate Medium is available as dehydrated culture medium or in ready-to-use prepared media tubes and bottles and in formats that meet international industrial guidelines for microbiological testing, including United States Pharmacopeia (USP) guidelines for sterility testing.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Certifications/ComplianceAOAC, BAM, COMPF, USP
ColorClear broth w/visible agar light to medium amber
FormatBottle
Manufacturing OriginUnited States
Media TypeFluid Thioglycollate Medium
Product TypeMicrobiology Media
Fill Volume100 mL
Includesw/ Indicator, Dextrose Medium
Packaging TypeGlass Bottle
Quantity10/Pk.
Unit SizeEach
Contents & Storage
2 to 25°C