Columbia CNA Agar with Sheep Blood Medium
Columbia CNA Agar with Sheep Blood Medium
Thermo Scientific™

Columbia CNA Agar with Sheep Blood Medium

Isolate Gram-positive organisms from clinical samples with this selective media traditionally used to identify staphylococci, streptococci, and S. pneumoniae.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatMedia TypeIncludes
R0132010/Pk.85mm MonoplateCNA with 5% Sheep BloodColistin, Nalidixic Acid
R01322100/Pk.85mm MonoplateCNA with 5% Sheep BloodColistin, Nalidixic Acid
Catalog number R01320
Price (USD)/ Each
-
Request A Quote
Quantity:
10/Pk.
Format:
85mm Monoplate
Media Type:
CNA with 5% Sheep Blood
Includes:
Colistin, Nalidixic Acid
Effectively isolate Gram-positive organisms from clinical samples using Thermo Scientific™ Columbia CNA Agar with Sheep Blood, a selective microbiological medium that includes colistin and nalidixic acid to inhibit gram-negative bacilli. Columbia CNA Agar has traditionally been used to identify staphylococci and streptococci. The addition of sheep blood to the medium allows distinct identification of S. pneumoniae through the production of clear alpha-haemolysis.
Columbia CNA Agar with Sheep Blood contains antibiotics to inhibit S. albus and Micrococcus species as well as Gram-positive and Gram-negative rods. It suppresses growth of Proteus, Klebsiella and Pseudomonas species while permitting unrestricted growth of S. aureus, haemolytic streptococci and enterococci. This medium enables important Gram-positive cocci to be recognised more readily and isolated easily from the mixed bacterial populations contained in many clinical specimens and foods.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
ColorOpaque red
Format85mm Monoplate
Manufacturing OriginUnited States
Media TypeCNA with 5% Sheep Blood
Product TypeAgar
Fill Volume18 mL
IncludesColistin, Nalidixic Acid
Packaging TypeFlow Wrapped - Single Layer
Quantity10/Pk.
Unit SizeEach
Contents & Storage
2°C to 8°C