ViewRNA™ ISH Cell 740 Module
Product Image
Invitrogen™
ViewRNA™ ISH Cell 740 Module
The ViewRNA ISH Cell 740 Module is designed to work in conjunction with both the ViewRNA ISH Cell Assay KitRead more
Have Questions?
Catalog number QVC0200
also known as QVC0200-00
Price (USD)/ 1 kit
550.00
-
Add to cart
Price (USD)/ 1 kit
550.00
Add to cart
ViewRNA™ ISH Cell 740 Module
Catalog numberQVC0200
Price (USD)/ 1 kit
550.00
-
Add to cart
The ViewRNA ISH Cell 740 Module is designed to work in conjunction with both the ViewRNA ISH Cell Assay Kit (Cat. No. QVC0001) and ViewRNA Cell Plus Assay Kit (88-19000-99). It contains the signal amplification and AlexaFluor 750-based detection components for visualization of target mRNA using adherent or suspension cells. AlexaFluor 750 can be visualized with a Cy7 filter set, with excitation/emission maxima of 749/775 nm.
The QuantiGene ViewRNA ISH Cell 740 Module is supplied with enough reagents to process 24 assays when using the 24-well chamber plate assay format and 144 assays when using 96-well optical-bottom plates. The kit is configured for processing a minimum of 6-wells per experiment when using the 24-well plate assay format. If fewer assays are processed per experiment, reagent shortages may occur.
Reported applications: Immunohistochemical staining
Note: We recommend the use of a strong antifade mountant, such as Prolong Glass Antifade Mountant (Cat. No. P36980), to minimize photobleaching.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Immunohistochemical Staining
Product TypeRNA In Situ Hybridization ISH Cell 740 Module
Quantity24 Assays
Unit Size1 kit
Contents & Storage
• PreAmp 10-740
• Amp 10-740
• Label Probe (LP 1-740)