You need to enable JavaScript to run this app.
Mouse Kidney Total RNA
Invitrogen™
Mouse Kidney Total RNA
Mouse Kidney Total RNA is a total RNA sample extracted from mouse renal tissue for use as a positive
control
Read more
Have Questions?
Catalog number
QS0635
Price (USD)
/ 10 µg
ERR#1200 - This item cannot be ordered online.
Mouse Kidney Total RNA is a total RNA sample extracted from mouse renal tissue for use as a positive control in QuantiGene assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Species
Mouse
Sample Type
RNA
For Use With (Application)
Nucleic Acid Quantitation
Product Type
Total RNA
Quantity
10 μg
Sample Source
Pooled, Normal Tissue
Shipping Condition
Dry Ice
Unit Size
10 µg