You need to enable JavaScript to run this app.
Human Spleen Total RNA
Invitrogen™
Human Spleen Total RNA
Human Spleen Total RNA is a total RNA sample extracted from human splenic tissue for use as a positive
control
Read more
Have Questions?
Catalog number
QS0627
Price (USD)
/ 10 µg
ERR#1200 - This item cannot be ordered online.
Human Spleen Total RNA is a total RNA sample extracted from human splenic tissue for use as a positive control in QuantiGene assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Species
Human
Sample Type
RNA
For Use With (Application)
Nucleic Acid Quantitation
Product Type
Total RNA
Quantity
10 μg
Sample Source
Pooled, Normal Tissue
Shipping Condition
Dry Ice
Unit Size
10 µg