Human Kidney Total RNA
Human Kidney Total RNA
Invitrogen™
Human Kidney Total RNA
Human Kidney Total RNA is a total RNA sample extracted from human renal tissue for use as a positive controlRead more
Have Questions?
Catalog number QS0616
Price (USD)/ 10 µg
ERR#1200 - This item cannot be ordered online.
Human Kidney Total RNA is a total RNA sample extracted from human renal tissue for use as a positive control in QuantiGene assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
SpeciesHuman
Sample TypeRNA
For Use With (Application)Nucleic Acid Quantitation
Product TypeTotal RNA
Quantity10 μg
Sample SourcePooled, Normal Tissue
Shipping ConditionDry Ice
Unit Size10 µg