You need to enable JavaScript to run this app.
Human Heart Total RNA
Invitrogen™
Human Heart Total RNA
Human Heart Total RNA is a total RNA sample extracted from cardiac tissue for use as a positive control
in
Read more
Have Questions?
Catalog number
QS0614
Price (USD)
/ 10 µg
ERR#1200 - This item cannot be ordered online.
Human Heart Total RNA is a total RNA sample extracted from cardiac tissue for use as a positive control in QuantiGene assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Species
Human
Sample Type
RNA
For Use With (Application)
Nucleic Acid Quantitation
Product Type
Total RNA
Quantity
10 μg
Sample Source
Pooled, Normal Tissue
Shipping Condition
Dry Ice
Unit Size
10 µg