You need to enable JavaScript to run this app.
T-cell Leukemia (Jurkat) Total RNA
Invitrogen™
T-cell Leukemia (Jurkat) Total RNA
T-Cell Leukemia (Jurkat) Total RNA is a total RNA sample extracted from Jurkat cells for use as a positive
control
Read more
Have Questions?
Catalog number
QS0601
Price (USD)
/ 10 µg
ERR#1200 - This item cannot be ordered online.
T-Cell Leukemia (Jurkat) Total RNA is a total RNA sample extracted from Jurkat cells for use as a positive control in QuantiGene assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Species
Human
Sample Type
RNA
For Use With (Application)
Nucleic Acid Quantitation
Product Type
Total RNA
Quantity
10 μg
Sample Source
Tumor or Diseased Tissue, Cell Line
Shipping Condition
Dry Ice
Unit Size
10 µg