QuantiGene™ Singleplex HT Assay Kit
QuantiGene™ Singleplex HT Assay Kit
Invitrogen™

QuantiGene™ Singleplex HT Assay Kit

QuantiGene Singleplex HT assays are hybridization-based assays for the direct quantitation of RNA, DNA, or microRNA transcripts directly from cellRead more
Have Questions?
Catalog number QS0384
Price (USD)/ 50 x 384-well plates
52,800.00
Add to cart
Price (USD)/ 50 x 384-well plates
52,800.00
Add to cart
QuantiGene™ Singleplex HT Assay Kit
Catalog numberQS0384
Price (USD)/ 50 x 384-well plates
52,800.00
-
Add to cart
QuantiGene Singleplex HT assays are hybridization-based assays for the direct quantitation of RNA, DNA, or microRNA transcripts directly from cell lysates or tissue homogenates. They utilize branched DNA technology for signal amplification and signal is detected using a standard luminometer. The QuantiGene Singleplex Assay Kit contains all buffers, plates, and detection reagents necessary to perform the assay in 384-well plates.

Additional plate seals are available for separate purchase (Cat. No. QS0506).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format384-well
For Use With (Application)QuantiGene Singleplex Probe Set
For Use With (Equipment)Luminometer
Product LineQuantiGene Singleplex
Product TypeSingleplex HT Assay Kit
Quantity50 Plates
Shipping ConditionBlue Ice
SpeciesUniversal
Sample TypePurified RNA, cell/blood/FFPE lysates, tissue homogenates
Unit Size50 x 384-well plates
Contents & Storage
Blocking Reagent (store at -20°C)
Pre-Amplifier (store at -20°C)
Amplifier (store at -20°C)
384-well Capture Plate (store at 2-8°C)
Label Probe (store at 2-8°C)
Substrate (store at 2-8°C)
Amplifier/Label Probe Diluent ( store at 15-30°C)
Lysis Mixture (store at 15-30°C)
Wash Buffer Component 1 (store at 15-30°C)
Wash Buffer Component 2 (store at 15-30°C)
Plate Seals (store at 15-30°C)