Contents & Storage
• Blocking Reagent (10 mL, store at 2–8°C)
• Pre-Amplifier Solution (570 mL, store at 2–8°C)
• Amplifier Solution(570 mL, store at 2–8°C)
• Capture Plate (50 ea, packaged individually, store at 2–8°C)
• Label Probe (550 μL, store at 2–8°C)
• Substrate (530 mL, store at 2–8°C)
• Label Probe Diluent (570 mL, store at 15–30°C)
• Lysis Mixture (410 mL, store at 15–30°C)
• Wash Buffer Component 1 (75 mL, store at 15–30°C)
• Wash Buffer Component 2 (125 mL, store at 15–30°C)
• Plate Seals Day 1 (50 ea, store at 15–30°C)
• Plate Seals Day 2 (210 ea, store at 15–30°C)