QuantiGene™ Plex 384-well Assay Kit
QuantiGene™ Plex 384-well Assay Kit
Invitrogen™

QuantiGene™ Plex 384-well Assay Kit

The QuantiGene Plex 384-well Assay Kit contains the generic reagents, buffers, and plates necessary to perform the assay in 384-wellRead more
Have Questions?
Catalog number QP1016
Price (USD)/ 1 kit
5,250.00
Add to cart
Price (USD)/ 1 kit
5,250.00
Add to cart
QuantiGene™ Plex 384-well Assay Kit
Catalog numberQP1016
Price (USD)/ 1 kit
5,250.00
-
Add to cart
The QuantiGene Plex 384-well Assay Kit contains the generic reagents, buffers, and plates necessary to perform the assay in 384-well plates and must be used with a target-specific QuantiGene Plex panel (sold separately).

See related sample preparation reagents and RNA controls ›

The QuantiGene Plex Assay is a gene expression assay that enables multiplexed measurement of up to 80 RNA transcript targets. The assay uses target-specific probes to capture the RNA of interest, and branched DNA technology then amplifies fluorescent signals, which are read with a Luminex xMAP instrument (FlexMap3D).

Features of the QuantiGene Plex 384-well Assay Kit include:
Accommodates difficult sample types—works with degraded and cross-linked RNA in FFPE tissues and blood
True multiplexing—measures up to 80 genes of interest and housekeeping genes in the same well with no cross-reactivity
Standardized platform—uses 384-well plate format compatible with Luminex FlexMap3D systems
Simple workflow—employs an ELISA-like workflow for direct hybridization of transcripts to beads and transcript labeling
Large inventory of validated genes—select from over 22,000 genes to create pathway and disease-themed panels
Rapid turnaround for custom panels—a custom panel can be created within three weeks

Technology
QuantiGene assays utilize branched DNA (bDNA) technology that has been used for decades in various applications and areas of biology. First, cells are lysed or tissue samples are homogenized to release the target RNA. Second, the oligonucleotide probe set is incubated with the target RNA and Luminex xMAP bead set overnight. During this incubation, the probes cooperatively hybridize to the target and capture probes bound to each bead, capturing the target RNA. Third, signal amplification is performed via sequential hybridization of the bDNA pre-amplifier, amplifier, and label probe molecules to the target. Addition of streptavidin phycoerythrin generates a luminescent signal directly proportional to the amount of target mRNA present in the sample.

Applications
QuantiGene Plex 384-well Assay applications include compound screening of drug candidates (primary screening), biomarker verification (secondary screening), siRNA knockdown efficiency, prospective and/or retrospective analysis of samples, microarray validation, predictive toxicology, and detection of translocations and fusion genes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format384-well
For Use With (Application)QuantiGene Plex Panel
For Use With (Equipment)Invitrogen™ Luminex™ FLEXMAP 3D™ Instrument System
Product LineQuantiGene Plex
Product TypeQuantiGene Plex 384-well Assay Kit
Quantity2 Plates
Shipping ConditionBlue Ice
SpeciesUniversal
Sample TypePurified RNA, cell/blood/FFPE lysates, tissue homogenates
Detection MethodFluorescence
Unit Size1 kit
Contents & Storage
• 200 μL Proteinase K, -20°C
• 2.1 mL Blocking Reagent, 2°C to 8°C
• 31 mL Label Probe Solution, 2°C to 8°C
• 31 mL Pre-Amplifier Solution, 2°C to 8°C
• 31 mL Amplifier Solution, 2°C to 8°C
• 280 μL SAPE, 2°C to 8°C
• 31 mL SAPE Diluent, 2°C to 8°C
• 24 mL Lysis Mixture, room temperature
• 6 mL Wash Buffer Component 1, room temperature
• 100 mL Wash Buffer Component 2, room temperature
• 1000 mL SAPE Wash Buffer, room temperature
• 2 Hybridization plates (Day 1) 384-well, room temperature
• 4 Manual Pressure seals (Day 1), room temperature
• 4 Heat seals (Day 1), room temperature
• 2 Magnetic Separation plates (Day 2), room temperature
• 10 Plates seals (Day 2), room temperature