The Qubit Flex System Verification Assay Kit is a fast, easy-to-use, reagent-based assay that tests the performance of the QubitRead more
Have Questions?
Catalog number Q33254
Price (USD)/ 25 assays
98.65
Online exclusive
100.00
Save 1.35 (1%)
-
Add to cart
Price (USD)/ 25 assays
98.65
Online exclusive
100.00
Save 1.35 (1%)
Add to cart
Qubit™ Flex System Verification Assay Kit
Catalog numberQ33254
Price (USD)/ 25 assays
98.65
Online exclusive
100.00
Save 1.35 (1%)
-
Add to cart
The Qubit Flex System Verification Assay Kit is a fast, easy-to-use, reagent-based assay that tests the performance of the Qubit Flex Fluorometer. The kit consists of three components: a blank reagent solution, a green fluorescent reagent, and a far-red fluorescent reagent. The green fluorescent reagent is designed to test blue excitation (470 nm LED; 430-495 nm excitation filter) and green fluorescence detection (510-580 nm emission filter). The far-red fluorescent reagent is designed to test red excitation (635 nm LED; 600-645 nm excitation filter) and far-red fluorescence detection (665-720 nm emission filter).
Note: The Qubit Flex System Verification Assay Kit can only be used with the Qubit Flex Fluorometer.
For Research Use Only. Not for use in diagnostic procedures.