Biotin-Anti-His-Tag Antibody
Biotin-Anti-His-Tag Antibody
Invitrogen™

Biotin-Anti-His-Tag Antibody

Biotin-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partiallyRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PV608925 μg
PV60901 mg
Catalog number PV6089
Price (USD)/ 25 µg
1,371.65
Online exclusive
1406.00 
Save 34.35 (2%)
-
Add to cart
Quantity:
25 μg
Price (USD)/ 25 µg
1,371.65
Online exclusive
1406.00 
Save 34.35 (2%)
Add to cart
Biotin-Anti-His-Tag Antibody
Catalog numberPV6089
Price (USD)/ 25 µg
1,371.65
Online exclusive
1406.00 
Save 34.35 (2%)
-
Add to cart
Biotin-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using biotin negatively impacts protein activity. These antibodies can be used with our Kinase Tracers and Eu-Streptavidin to conduct LanthaScreen™ Eu Kinase Binding Assays. Learn More and See Which Kinases Have Been Validated Using This Assay
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ApplicationsKinase Assay, Kinase Assay
Concentration0.5 mg⁄ml
ConjugateBiotin
DescriptionBiotin-Anti-His-Tag Antibody, 25μg, 0.5mg/mL Concentration, HIT2 Clone, 150kDa Molecular Weight, Biotin Conjugate, Used For Biochemical Kinase Assays, Drug Discovery and Development, Kinase Biology, LanthaScreen™ kinase binding assays
Molecular Weight of Antigen150kDa
Quantity25 μg
Regulatory StatusRUO, RUO
Research DisciplineKinase Biology
Primary or SecondaryPrimary
Packaging1 Each
FormFrozen
Assay EntryBiochemical binding/kinetic
Shipping ConditionDry Ice
Target EntryABL1 G250E, STK16, ZAP70, MAP2K1 S218D S222D, MAPK8, CAMK2B, CHEK1, STK33, IGFR1, BLK, LYNA, NEK2, CHUK, MAPKAPK5, MAP3K7/MAP3K7IP1, DAPK2, HIPK3, MAP2K1, MAP2K6, PIK3CG, ABL1 E255K, BMX, MAP4K2, MAPK9, CAMK1D, CHEK2, PIM1, STK22D, CSNK1G2, EPHA3, EPHB4, LCK, YES1, MET, PRKCI, WNK3, ABL1, ABL2, FGFR3, FGFR3 K650E, KIT V654A, AURKB, CDC42BPA, CDK1, CDK8, CDK9/Cyclin K, LYNB, CDC42BPB, CDK2, CDK7, PDGFRB, MET M1250T, RPS6KA1, LIMK1, MAP2K2, PIK3CA/PIK3R1, MAP2K6 S207E T211E, SRC, ABL1 Y253F, KIT T670I, RPS6KA2, PAK7, IRAK4, CSK
Unit Size25 µg
Contents & Storage
Biotin-labeled anti-His tag antibody in HEPES buffered saline. Store at -20°C.