LanthaScreen™ Eu-anti-DYKDDDDK Antibody Kit
Product Image
Thermo Scientific™

LanthaScreen™ Eu-anti-DYKDDDDK Antibody Kit

LanthaScreen™ Europium-labeled anti-DYKDDDDK antibodies are used to indirectly label proteins tagged with DYKDDDDK (Flag). This can be advantageous when workingRead more
Have Questions?
Catalog number PV6034
Price (USD)/ 1 kit
373.00
-
Add to cart
Price (USD)/ 1 kit
373.00
Add to cart
LanthaScreen™ Eu-anti-DYKDDDDK Antibody Kit
Catalog numberPV6034
Price (USD)/ 1 kit
373.00
-
Add to cart

LanthaScreen™ Europium-labeled anti-DYKDDDDK antibodies are used to indirectly label proteins tagged with DYKDDDDK (Flag). This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.25 mg/mL
ConjugateEuropium/Lanthanide Chelates
Quantity1 Kit
FormFrozen
Excitation/Emission340/615, 665 nm
Shipping ConditionDry Ice
Product LineLanthaScreen™
Unit Size1 kit
Contents & Storage
Europium-labeled anti-DYKDDDDK antibody in HEPES buffered saline. Store at -20°C.