LanthaScreen™ Eu-Streptavidin Kit
Product Image

LanthaScreen™ Eu-Streptavidin Kit

A general reagent for use in assay development, the Europium-Streptavidin binds to biotinylated peptide or physiological protein substrates. The Eu-streptavidinRead more
Have Questions?
Catalog number PV6030
Price (USD)/ 1 kit
415.00
-
Add to cart
Price (USD)/ 1 kit
415.00
Add to cart
LanthaScreen™ Eu-Streptavidin Kit
Catalog numberPV6030
Price (USD)/ 1 kit
415.00
-
Add to cart
A general reagent for use in assay development, the Europium-Streptavidin binds to biotinylated peptide or physiological protein substrates. The Eu-streptavidin Kit contains 10 μg Eu-Streptavidin, 400 μl AlexaFluor® 647-biotin control peptide, and 100 mL of TR-FRET Dilution Buffer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.25 mg/mL
ConjugateAlexa Fluor 647, Europium
Quantity1 kit
Excitation/Emission340/615, 665
Shipping ConditionDry Ice
FormLiquid
Product LineLanthaScreen™
BufferTR-FRET Dilution Buffer
Product TypeStreptavidin Conjugate (Fluorescent)
Label or DyeAlexa Fluor 647, Europium
Unit Size1 kit
Contents & Storage
The Eu-Streptavidin Kit contains 10 μg Eu-streptavidin, 400 μl AlexaFluor™ 647-biotin control peptide and 100 mL of TR-FRET Dilution Buffer. Store reagents as indicated on the certificate of analysis.