LanthaScreen Europium-labeled anti-DYKDDDDK antibodies are used to indirectly label proteins tagged with DYKDDDDK (Flag). This can be advantageous when workingRead more
Have Questions?
Change view
Catalog Number
Quantity
PV6027
1 mg
PV6026
25 μg
2 Options
Catalog number PV6027
Price (USD)/ 1 mg
13,670.00
-
Add to cart
Quantity:
1 mg
Price (USD)/ 1 mg
13,670.00
Add to cart
LanthaScreen™ Eu-anti-DYKDDDDK Antibody
Catalog numberPV6027
Price (USD)/ 1 mg
13,670.00
-
Add to cart
LanthaScreen Europium-labeled anti-DYKDDDDK antibodies are used to indirectly label proteins tagged with DYKDDDDK (Flag). This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.25 mg/mL
ConjugateEuropium/Lanthanide Chelates
Quantity1 mg
FormFrozen
Excitation/Emission340/615, 665
Shipping ConditionDry Ice
Product LineLanthaScreen™
Unit Size1 mg
Contents & Storage
Europium-labeled anti-DYKDDDDK antibody in HEPES buffered saline. Store at -20°C.