LanthaScreen™ Eu-anti-DYKDDDDK Antibody
LanthaScreen™ Eu-anti-DYKDDDDK Antibody

LanthaScreen™ Eu-anti-DYKDDDDK Antibody

LanthaScreen Europium-labeled anti-DYKDDDDK antibodies are used to indirectly label proteins tagged with DYKDDDDK (Flag). This can be advantageous when workingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PV602625 μg
PV60271 mg
Catalog number PV6026
Price (USD)/ 25 µg
1,174.00
-
Add to cart
Quantity:
25 μg
Price (USD)/ 25 µg
1,174.00
Add to cart
LanthaScreen™ Eu-anti-DYKDDDDK Antibody
Catalog numberPV6026
Price (USD)/ 25 µg
1,174.00
-
Add to cart
LanthaScreen Europium-labeled anti-DYKDDDDK antibodies are used to indirectly label proteins tagged with DYKDDDDK (Flag). This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.25 mg⁄ml
ConjugateAlexa Fluor 647/Europium/Lanthanide Chelates
FormulationFrozen
Quantity25 μg
Regulatory StatusRUO, RUO
Research DisciplineTarget and Lead Identification and Validation
Primary or SecondaryPrimary
Packaging1 Each
FormFrozen
Excitation/Emission340⁄615, 665
Shipping ConditionDry Ice
Product LineLanthaScreen™
Unit Size25 µg
Contents & Storage
Europium-labeled anti-DYKDDDDK antibody in HEPES buffered saline. Store at -20°C.