LanthaScreen™ Eu-Streptavidin
Product Image
Invitrogen™

LanthaScreen™ Eu-Streptavidin

A general reagent for use in assay development, the Europium-Streptavidin binds to biotinylated peptide or physiological protein substrates.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PV589925 μg
PV60251 mg
Catalog number PV5899
Price (USD)/ 25 µg
1,038.65
Online exclusive
1104.00 
Save 65.35 (6%)
-
Add to cart
Quantity:
25 μg
Price (USD)/ 25 µg
1,038.65
Online exclusive
1104.00 
Save 65.35 (6%)
Add to cart
LanthaScreen™ Eu-Streptavidin
Catalog numberPV5899
Price (USD)/ 25 µg
1,038.65
Online exclusive
1104.00 
Save 65.35 (6%)
-
Add to cart

A general reagent for use in assay development, the Europium-Streptavidin binds to biotinylated peptide or physiological protein substrates.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConcentrationSee the Certificate of Analysis
ConjugateAlexa Fluor 647, Europium
Quantity25 μg
Assay EntryBiochemical binding/kinetic
Excitation/Emission340/615, 665 nm
Shipping ConditionDry Ice
Target EntryABL1 G250E, STK16, ZAP70, MAP2K1 S218D S222D, MAPK8, CAMK2B, CHEK1, STK33, IGFR1, BLK, LYNA, NEK2, CHUK, MAPKAPK5, MAP3K7/MAP3K7IP1, DAPK2, HIPK3, MAP2K1, MAP2K6, PIK3CG, ABL1 E255K, BMX, MAP4K2, MAPK9, CAMK1D, CHEK2, PIM1, STK22D, CSNK1G2, EPHA3, EPHB4, LCK, YES1, MET, PRKCI, WNK3, ABL1, ABL2, FGFR3, FGFR3 K650E, KIT V654A, AURKB, CDC42BPA, CDK1, CDK8, CDK9/Cyclin K, LYNB, CDC42BPB, CDK2, CDK7, PDGFRB, MET M1250T, RPS6KA1, LIMK1, MAP2K2, PIK3CA/PIK3R1, MAP2K6 S207E T211E, SRC, ABL1 Y253F, KIT T670I, RPS6KA2, PAK7, IRAK4, CSK
FormLiquid
Product LineLanthaScreen™
Product TypeStreptavidin Conjugate (Fluorescent)
Label or DyeAlexa Fluor 647, Europium
Unit Size25 µg
Contents & Storage
Europium-labeled Streptavidin in HEPES buffered saline. Store at -20°C.