LanthaScreen™ Eu-anti-His Tag Antibody Kit
LanthaScreen™ Eu-anti-His Tag Antibody Kit

LanthaScreen™ Eu-anti-His Tag Antibody Kit

LanthaScreen™ europium-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working withRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PV559625 μg
PV55971 mg
Catalog number PV5596
Price (USD)/ 25 µg
1,476.00
-
Add to cart
Quantity:
25 μg
Price (USD)/ 25 µg
1,476.00
Add to cart
LanthaScreen™ Eu-anti-His Tag Antibody Kit
Catalog numberPV5596
Price (USD)/ 25 µg
1,476.00
-
Add to cart
LanthaScreen™ europium-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity. These antibodies can be used with our Kinase Tracers to conduct LanthaScreen™ Eu Kinase Binding Assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AntigenHis
ApplicationsKinase Assay, Kinase Assay
Concentration0.25 mg/mL, 0.25 mg/ml, 0.25 mg⁄ml
ConjugateEuropium
Molecular Weight of Antigen150kDa
Quantity25 μg
Regulatory StatusRUO, RUO
Research DisciplineKinase Biology
Primary or SecondaryPrimary
Packaging1 Kit(s)
Antibody Molecular Weight150 kDa
FormLiquid
Assay EntryBiochemical binding/kinetic
Excitation/Emission340⁄665
For Use With (Equipment)Microplate Reader
Shipping ConditionDry Ice
Target EntryCSF1R, PRKCH, PRKCQ, GSK3B, FGFR1, TEC, CAMK2D, RIPK2, FYN, AURKA, NUAK1, FLT3, KDR, CAMK2A, RPS6KA3, AXL, FGR, CAMKK1, CDK9/Cyclin T1, TBK1, PAK6, PDK1, FLT3 D835Y, BTK, FES, PRKACA, PTK6
Target MoleculeHis
TechniqueTR-FRET
Product LineLanthaScreen™
Unit Size25 µg
Contents & Storage
Europium-labeled 6×His antibody is supplied in HEPES-buffered saline. Store at -20°C.