LanthaScreen™ TR-FRET PXR (SXR) Competitive Binding Assay Kit, goat
Product Image

LanthaScreen™ TR-FRET PXR (SXR) Competitive Binding Assay Kit, goat

This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15142:The LanthaScreen™ TR-FRET PXR (SXR) CompetitiveRead more
Have Questions?
Catalog number PV4839
Price (USD)/ 800 x 20 µL assays
2,435.00
-
Add to cart
Price (USD)/ 800 x 20 µL assays
2,435.00
Add to cart
LanthaScreen™ TR-FRET PXR (SXR) Competitive Binding Assay Kit, goat
Catalog numberPV4839
Price (USD)/ 800 x 20 µL assays
2,435.00
-
Add to cart
This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15142:

The LanthaScreen™ TR-FRET PXR (SXR) Competitive Binding Assay Kit provides a sensitive and robust method for high-throughput screening (HTS) of ligands for the pregnane X receptor (PXR), also known as the steroid and xenobiotic receptor (SXR). The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescent small-molecule PXR ligand (Fluormone™ PXR (SXR) Green), and a human PXR (SXR) ligand-binding domain that is tagged with glutathione S-transferase (GST) in a homogeneous mix-and-read assay format.

To assay:
When running the LanthaScreen™ TR-FRET PXR (SXR) competitive binding assay, a Tb-labeled anti-GST antibody is used to indirectly label the receptor by binding to its GST tag. Competitive ligand binding is detected by a test compound’s ability to displace a fluorescent ligand (tracer) from the receptor, which results in a loss of FRET signal between the Tb-anti-GST antibody and the tracer (Figure 1). This type of binding assay is analogous to radioligand-based assays, except that it eliminates handling of radioactivity and enables a homogeneous, “addition-only ” format.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateTb (Terbium)
Detection MethodFluorescence
For Use With (Equipment)Microplate Reader
Packaging384-well Plate
Assay EntryBiochemical competitive binding
Gene ID (Entrez)8856
ReadoutEnd Point
Shipping ConditionDry Ice
Target EntryNR1I2, PXR(SXR)
Product LineLanthaScreen™
For Use With (Application)Competitive Binding Assay, TR-FRET
LigandPXR
Product TypeTR-FRET PXR (SXR) Competitive Binding Assay Kit
No. of Assays800 x 20 μL assays
Unit Size800 x 20 µL assays
Contents & Storage
Store in ultra-cold freezer (-68 to -85°C).