LanthaScreen™ TR-FRET ERR beta Coactivator Assay Kit, goat
Product Image
Thermo Scientific™

LanthaScreen™ TR-FRET ERR beta Coactivator Assay Kit, goat

This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15139:The LanthaScreen™ TR-FRET ERR (estrogen relatedRead more
Have Questions?
Catalog number PV4800
Price (USD)/ 800 x 20 µL assays
2,175.00
-
Add to cart
Price (USD)/ 800 x 20 µL assays
2,175.00
Add to cart
LanthaScreen™ TR-FRET ERR beta Coactivator Assay Kit, goat
Catalog numberPV4800
Price (USD)/ 800 x 20 µL assays
2,175.00
-
Add to cart

This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15139:

The LanthaScreen™ TR-FRET ERR (estrogen related receptor) beta Coactivator Assay Kit provides a sensitive and robust method for highthroughput screening of potential ERR beta ligands as inverse agonists of ligand-dependent coactivator displacement. The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and a human recombinant ERR beta ligand-binding domain (ERR beta-LBD) that is tagged with glutathione-S-transferase (GST) in a homogeneous mix-and-read assay format.

To assay:

When running the LanthaScreen™ ERR beta TR-FRET Coactivator Assay, ERR beta-LBD is added to ligand test compounds followed by addition of a mixture of the fluorescein-coactivator peptide and Tb-labeled anti-GST antibody. After an incubation period at room temperature, the 520 nm/495 nm TR-FRET ratio is calculated and can be used to determine the EC50 from a dose response curve of the compound.

Based on the biology of the ERR beta-coactivator peptide interaction, this ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and displace coactivator peptide (see Figure 1).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateTb (Terbium)
Detection MethodFluorescence
For Use With (Equipment)Microplate Reader
Packaging384-well Plate
Assay EntryBiochemical coregulator interaction
ReadoutEnd Point
Shipping ConditionDry Ice
Target EntryNR3B2, ERR2, ESRRB
Product LineLanthaScreen™
For Use With (Application)Co-Factor Interaction Assay, TR-FRET
LigandERR beta
Product TypeTR-FRET ERR Beta Coactivator Assay Kit
No. of Assays800 x 20 μL Assays
Unit Size800 x 20 µL assays
Contents & Storage
The LanthaScreen™ TR-FRET ERR beta Coactivator Assay Kit contains ERR beta-LBD (GST) protein, fluorescently labeled PGC1a coactivator peptide, Tb-labeled anti-GST antibody, and buffers.

Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).